missing translation for 'onlineSavingsMsg'
Learn More

PHLPPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16803825 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16803825 20 μL 20µL
16893815 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16803825

Brand: Proteintech 252441AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Protein phosphatase involved in regulation of Akt and PKC signaling. Mediates dephosphorylation in the C-terminal domain hydrophobic motif of members of the AGC Ser/Thr protein kinase family; specifically acts on ′Ser-473′ of AKT1, ′Ser-660′ of PRKCB isoform beta-II and ′Ser-657′ of PRKCA. Akt regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes. Dephosphorylation of ′Ser-473′ of Akt triggers apoptosis and decreases cell proliferation. Also controls the phosphorylation of AKT3. Dephosphorylates STK4 on ′Thr-387′ leading to STK4 activation and apoptosis. Dephosphorylates RPS6KB1 and is involved in regulation of cap-dependent translation. Inhibits cancer cell proliferation and may act as a tumor suppressor. Dephosphorylation of PRKCA and PRKCB leads to their destabilization and degradation. Dephosphorylates RAF1 inhibiting its kinase activity.

Specifications

Antigen PHLPPL
Applications Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PHLPP2
Gene Accession No. Q6ZVD8, Q8BXA7
Gene Alias KIAA0931, PHLPP like, PHLPP2, PHLPPL
Gene Symbols Phlpp2
Host Species Rabbit
Immunogen PHLPPL Fusion Protein Ag19000
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23035, 244650, 498949
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.