Learn More
Phloroglucinol, Sigma-Aldrich™
Specifications
Specifications
| CAS | 108-73-6 |
| Melting Point | 217°C to 220°C |
| Molecular Formula | C6H3-1,3,5-(OH)3 |
| MDL Number | MFCD00002286 |
| Quantity | 100 g |
| Synonym | Phloroglucinol, 1,3,5-Trihydroxybenzene, Phloroglucinol |
| Molecular Weight (g/mol) | 126.11 |
| Grade | Research |