Learn More
Thermo Scientific™ Phire Reaction Buffers
Product Code. 13515170 Shop All Thermo Scientific Products
Change view
Click to view available options
Chemical Name or Material:
Phire Green Reaction Buffer
Phire Reaction Buffer, detergent-free
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Name or Material Unit Size
13515170 Phire Green Reaction Buffer Pack of 4
10525709 Phire Reaction Buffer, detergent-free Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13515170 Supplier Thermo Scientific™ Supplier No. F527L
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Includes
Phire Green Reaction Buffer
  • 5x Phire Green Reaction Buffer (1X buffer provides 1.5 mM MgCl2)
  • 50 mM MgCl2 solution
  • DMSO
The reaction buffer for Thermo Scientific Phire Hot Start II DNA Polymerase is available in three formats: non-colored version with and without detergents or Green version containing a density reagent and two tracking dyes for direct loading of PCR products on gel.
Thermo Scientific Phire Reaction Buffer is designed for Phire Hot Start II DNA Polymerase. This 5X buffer solution is available in three formats, a non-colored version with or without detergents and a green version that contains a density reagent and two tracking dyes for direct loading of PCR products on gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.
Phire Reaction Green Buffer includes a blue die that migrates with 3–5 kb DNA fragments in 1% agarose gel and a yellow dye that migrates faster than 10 bp DNA fragments in 1% agarose gel. The dyes have excitation peaks at 424 nm and 615 nm.
The concentration of Mg2+ in Phire Reaction Buffer is optimized to work well for most amplicons. The buffer contains 1.5 mM MgCl2 at final reaction concentration. Separate tubes of MgCl2 and DMSO are provided for further optimization.
Notes
  • For applications that require PCR product analysis by absorbance or fluorescence excitation, the colorless Phire Reaction Buffer is recommended.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phire Green Reaction Buffer
Description Green
Concentration 5X
For Use With (Application) Hot-start PCR
Quantity 4 x 1.5 mL
Includes • 5X Phire Green Reaction Buffer, 4 x 1.5 mL
• 50 mM MgCl2, 2 x 1.5 mL
• DMSO, 0.5 mL
Store at -20°C.