missing translation for 'onlineSavingsMsg'
Learn More

PhiKan 083, MedChemExpress

Product Code. p-200098255
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. Quantity unitSize
16462996 25 mg 25mg
16446937 5 mg 5mg
16434218 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16462996

Brand: Medchem Express HY108637

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PhiKan 083 is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM. PhiKan 083 can be used for cancer research.

PhiKan 083 is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM. PhiKan 083 can be used for cancer research.

Specifications

Chemical Name or Material PhiKan 083
CAS 880813-36-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H18N2
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (419.59 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (419.59 mM; Need ultrasonic)
SMILES CNCC1=CC2=C(C=C1)N(CC)C3=C2C=CC=C3
Molecular Weight (g/mol) 238.33
Formula Weight 238.33
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.