missing translation for 'onlineSavingsMsg'
Learn More

PHGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14719-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16873504

  • 4593.76 NOK / 150µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane.
Specifications

Specifications

PHGDH
Polyclonal
Unconjugated
PHGDH
3 PGDH, 3PGDH, PDG, PGAD, PGD, PGDH, PGDH3, PHGDH, phosphoglycerate dehydrogenase, SERA
Rabbit
Antigen Affinity Chromatography
RUO
236539, 26227, 58835
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08651, O43175, Q61753
PHGDH
PHGDH Fusion Protein Ag6445
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers