missing translation for 'onlineSavingsMsg'
Learn More

PHF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16836345 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16836345 150 μL 150µL
16846345 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16836345

Brand: Proteintech 510621AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

May be involved in the regulation of serine proteinases present in the brain or extravasated from the blood. Inhibitor of cathepsin G, kallikrein-8 and thrombin. May play an important role in the inner ear in the protection against leakage of lysosomal content during stress and loss of this protection results in cell death and sensorineural hearing loss.

Specifications

Antigen PHF6
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PHF6
Gene Accession No. Q8IWS0, Q9D4J7
Gene Alias BORJ, KIAA1823, PHD finger protein 6, PHD like zinc finger protein, PHF6
Gene Symbols Phf6
Host Species Rabbit
Immunogen PHF6 Fusion Protein Ag2102
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70998, 84295
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.