Learn More
PHF5A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15554-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as a transcriptional regulator by binding to the GJA1/Cx43 promoter and enhancing its up-regulation by ESR1/ER- alpha. Also involved in pre-mRNA splicing.Specifications
| PHF5A | |
| Polyclonal | |
| Unconjugated | |
| PHF5A | |
| bK223H9.2, INI, PHD finger protein 5A, PHF5A, SF3b14b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 68479, 84844 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P83870, Q7RTV0 | |
| PHF5A | |
| PHF5A Fusion Protein Ag7918 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |