missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF15 Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11513-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
PHF15 | |
Polyclonal | |
Unconjugated | |
JADE2 | |
JADE2, KIAA0239, PHD finger protein 15, PHF15, Protein Jade 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23338, 481508 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NQC1 | |
JADE2 | |
PHF15 Fusion Protein Ag2073 | |
150 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG |