Learn More
PHEX Mouse anti-Human, Unconjugated, Clone: 1D4B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66934-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The PHEX antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PHEX. This antibody recognizes human, rat, pig antigen. The PHEX antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PHEX | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Pig |
Western Blot, Immunohistochemistry, ELISA | |
1D4B2 | |
Unconjugated | |
20 μL | |
5251 | |
IgG1 |