Learn More
PHEX Mouse anti-Human, Unconjugated, Clone: 1D4B2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66934-1-IG-20UL
 View more versions of this product
Product Code. 16809245
  • 1710.33 NOK / 20µL
Description
The PHEX antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PHEX. This antibody recognizes human, rat, pig antigen. The PHEX antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PHEX
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
1D4B2
Unconjugated
20 μL
5251
IgG1