Learn More
uvex Pheos Eyewear
Product Code. 17757689 Shop All Uvex Products
Change view
Click to view available options
Frame Color: Black and gray ; Coating: supravision ETC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17757689 Frame Color: Black and gray ; Coating: supravision ETC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17757689 Supplier uvex Supplier No. 9192415
  • ONLINE EXCLUSIVE PRICE
    161.00 NOK
    154.73 NOK / Pack of 1
    Save 6.27 NOK
    3% Off
Estimated Shipment: 08-12-2026
More than just a common spectacle - the modern fashion-look makes it a welcome companion at everyday work
  • Perfect all-round protection, large field of vision through duo spherical lens, outstanding ventilation thanks to integrated uvex climazone temple concept
  • Coating uvex supravision excellence
  • TRUSTED_SUSTAINABILITY
    Specifications
    Lens Type Supravision ETC
    Lens Coating Anti-fog
    Color Anthracite, Blue
    Certifications/Compliance (Europe) EN 166, EN 170
    Lens Tint Clear
    Product Type Ergonomic Safety Glasses
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Plastic
    UV Protection UV400
    Frame Type Standard