Description
Optimal eye protection in many situations: the soft browguard component is injection moulded onto the lens and keeps dust and liquid splashes out of the eyes. The innovative uvex x-tended eyeshield adapts perfectly to the wearer – for high levels of comfort and protection. The innovative IR-ex infrared protection filter provides reliable protection against infrared radiation while also enabling optimum recognition of signal colors.
- Sporty safety spectacles with high wearer acceptance
- Robust polycarbonate lens with proven uvex infradur coating technology – permanently anti-fog and scratch-resistant lens
- Metal free
- Frame color: black, red
- PC lens: grey IR-ex 5
- X-tended eyeshield: soft-seal frame technology wraps around the forehead and provides protection against particle penetration
- Ergonomically shaped arms with soft ear-pieces ensure a comfortable and secure fit without creating pressure points
- Soft, flexible nose piece adapts to the wearer, delivering a non-slip and pressure-free fit
- Certified in line with EN 166 (personal eye protection), EN 169 (filters for welding), EN 170 (UV filters) and EN 171 (infrared filters)
- Certification: W 166 FT CE – 4C-5 W1 FTKN CE, 2C-5 W1 FTKN CE
- UV400 – 100% protection against harmful UV radiation to 400 nm
Specifications
Specifications
| Lens Coating | Uncoated |
| Lens Tint | Clear |
| Product Type | Glasses (Standard) |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
| UV Protection | UV400 |
| Frame Type | Classic |
| Autoclavable | Not Autoclavable |