missing translation for 'onlineSavingsMsg'
Learn More

(Phenylsulfonyl)acetic acid, 97%

Product Code. 11388085
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11388085

Brand: Thermo Scientific Alfa Aesar L08299.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3959-23-7 | C8H8O4S | 200.208 g/mol

(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3959-23-7
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00025042
InChI Key YTEFAALYDTWTLB-UHFFFAOYSA-N
Synonym phenylsulfonyl acetic acid, 2-phenylsulfonyl acetic acid, 2-benzenesulfonyl acetic acid, phenylsulfonylacetic acid, acetic acid, phenylsulfonyl, benzenesulfonyl-acetic acid, benzenesulfonyl acetic acid, 2-phenylsulphonyl acetic acid, benzenesulfonylacetic acid, phenyl sulfonylacetic acid
PubChem CID 59543
IUPAC Name 2-(benzenesulfonyl)acetic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)CC(=O)O

Specifications

Melting Point 109°C to 114°C
Quantity 5 g
Beilstein 2504376
Formula Weight 200.22
Percent Purity 97%
Chemical Name or Material (Phenylsulfonyl)acetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.