Learn More
Phenylhydrazine, Sigma-Aldrich™
Product Code. 15869440
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869440 1 L 1 L
15849440 5 mL 5mL
15859440 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15869440 Supplier Sigma Aldrich Supplier No. 8072501000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-63-0
Specifications
Melting Point 19.6°C
Density 1.10 g/cm3 at 20°C
Boiling Point 244°C/1013 hPa
MDL Number MFCD00007573
Quantity 1 L
Synonym Phenylhydrazine, Phenylhydrazine
Molecular Weight (g/mol) 108.14
Grade Research