Learn More
Cytiva Phenyl Sepharose™ 6 Fast Flow Chromatography Media
Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
200 mL
Product Type:
Phenyl Sepharose 6 Fast Flow Media (high sub)
Phenyl Sepharose 6 Fast Flow Media (low sub)
Unit Size:
200mL
25mL
Each
Description
- Base matrix derivatized via uncharged, chemically-stable ether linkages
- Two levels of substitution help find optimal selectivity and binding capacity for given application
- Hydrophilic nature of the base matrix ensures low levels of non-specific binding
- Hydrophobic interaction chromatography (HIC) media composed of cross-linked 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
- Degree of substitution for low sub HIC media is ∽25μmol phenyl/mL gel
- Degree of substitution for high sub HIC media is ∽40μmol phenyl/mL gel
Specifications
Specifications
| Quantity | 25 mL |
| Stability | Stable in commonly used aqueous Buffers: 1 M NaOH, 3 M Ammonium Sulphate, 30% Isopropanol, 70% Ethanol, 10% Ethylene Glycol, 0.5% SDS, 6 M Guanidine-Hydrochloride, 8 M Urea |
| For Use With (Application) | For Capture and Intermediate PurifiCatIon Requiring Low to Medium Hydrophobicity |
| Storage Requirements | 4°C to 30°C |
| Product Type | Phenyl Sepharose 6 Fast Flow Media (low sub) |
| Flow Rate | 250 to 400 cm/Hoμr |
| Particle Size | 45 to 165 μm |
| Matrix | 6% Cross-Linked Agarose |
| pH Range | 3 to 13 |
| Storage Buffer | 20% Ethanol |
| Show More |
Safety and Handling
Product Identifier
- Phenyl Sepharose 6 Fast Flow (low sub)
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol