missing translation for 'onlineSavingsMsg'
Learn More

Phenyl[2-phenylthio)phenyl]carbamate, TRC

Product Code. 30427052
Change view
Click to view available options
Quantity:
500 mg
5 g
Product Code. Quantity
30427052 500 mg
30425713 5 g
2 options
This item is not returnable. View return policy

Product Code. 30427052

Brand: Toronto Research Chemicals TRCP335965500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111974-73-3
Formula Weight 321.0823
InChI Formula InChI=1S/C19H15NO2S/c21-19(22-15-9-3-1-4-10-15)20-17-13-7-8-14-18(17)23-16-11-5-2-6-12-16/h1-14H,(H,20,21)
IUPAC Name phenyl N-(2-phenylsulfanylphenyl)carbamate
Molecular Formula C19 H15 N O2 S
Molecular Weight (g/mol) 321.39
SMILES O=C(Nc1ccccc1Sc2ccccc2)Oc3ccccc3
Synonym Carbamic acid, N-[2-(phenylthio)phenyl]-, phenyl ester, Carbamic acid, [2-(phenylthio)phenyl]-, phenyl ester (9CI), Phenyl [2-(phenylthio)phenyl]carbamate, Phenyl N-[(2-Phenylsulfanyl)phenyl]carbamate, N-[2-(Phenylthio)phenyl]carbamic Acid Phenyl Ester
Chemical Name or Material Phenyl[(2-phenylthio)phenyl]carbamate

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.