missing translation for 'onlineSavingsMsg'
Learn More

Phenoxybenzamine hydrochloride, MedChemExpress

Product Code. p-200097734
Encompass_Preferred
Click to view available options
Quantity:
200 mg
500 mg
1 g
Unit Size:
1g
200mg
500mg
This item is not returnable. View return policy

Product Code. 16461416

Brand: Medchem Express HYB0431A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist that is commonly used for the research of hypertension, specifically caused by pheochromocytoma. Phenoxybenzamine hydrochloride also shows antitumor activity.

Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist that is commonly used for the research of hypertension, specifically caused by pheochromocytoma. Phenoxybenzamine hydrochloride also shows antitumor activity.

Specifications

Chemical Name or Material Phenoxybenzamine hydrochloride
CAS 63-92-3
Color White
Purity Grade Notes Research
Molecular Formula C18H23Cl2NO
Quantity 200 mg
Solubility Information DMSO : 100 mg/mL (293.87 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.97 mM; Need ultrasonic)
SMILES CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl
Molecular Weight (g/mol) 340.29
Formula Weight 340.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.