Learn More
Phenoxazine, 98%
Description
Phenoxazine is used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenoxazine is used as primary and secondary intermediates.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
Spécification
Spécification
| Melting Point | 154°C to 158°C |
| Quantity | 1 g |
| Beilstein | 143234 |
| Merck Index | 14,7253 |
| Solubility Information | Soluble in methanol. Insoluble in water. |
| Formula Weight | 183.21 |
| Percent Purity | 98% |
| Chemical Name or Material | Phenoxazine |