missing translation for 'onlineSavingsMsg'
Learn More

Phenothiazine, Sigma-Aldrich™

Product Code. 15871301
Change view
Click to view available options
Quantity:
100 g
1 kg
50 kg
Unit Size:
100g
1kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871301 50 kg 50kg
15851301 100 g 100g
15861301 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15871301 Supplier Sigma Aldrich Supplier No. 8209759050
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-84-2

Specifications

Melting Point 182°C to 185°C
Density 1.296 g/cm3 at 20°C
Boiling Point 381°C/1011 hPa
MDL Number MFCD00005015
Quantity 50 kg
Synonym Phenothiazine, Phenothiazine
Molecular Weight (g/mol) 199.27
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.