Learn More
Phenosafranine, Pure, C.I.50200, Fisher Chemical
Product Code. 10665882 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665882 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10665882 Supplier Fisher Chemical Supplier No. P/2450/44
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Specifica
CAS 81-93-6
Melting Point >300°C
Color Green
Packaging Glass Jar
Molecular Formula C18H15ClN4
MDL Number MFCD00036335
Quantity 5 g
Synonym phenosafranin, phenosafranine, phenosafranine, chloride, phenosafrinine, phenosofranine, phenosofrinine, safranin b extra, safrinin b extra, 3,7-diamino-5-phenylphenazinium chloride, schultz no. 958
InChI Key SOUHUMACVWVDME-UHFFFAOYSA-N
SMILES [Cl-].NC1=CC2=[N+](C3=CC=CC=C3)C3=CC(N)=CC=C3N=C2C=C1
IUPAC Name 10-phenylphenazin-10-ium-2,8-diamine;chloride
Molecular Weight (g/mol) 322.80
PubChem CID 65732
ChEBI CHEBI:33601
Formula Weight 322.8g/mol
Physical Form Powder
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Phenosafranin
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.