Learn More
Phenolphtalein Indicator In Propanol-2, Chem Lab™
Product Code. 11900048
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity unitSize
11900048 1 L 1 L
1 options
Product Code. 11900048
Brand: Chem-Lab CL06.0603.1000
Specifications
Melting Point -89°C
Odor Odorless
Packaging 1L
Color Colorless
pH <2
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenolphtalein 10 g/l in propanol-2 indicator solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iso-Propanol, Phenolphtalein indicator