missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, Supelco™

Product Code. 17168065
Change view
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17168065 100 g 100g
15848110 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17168065 Supplier Supelco Supplier No. 1593750100
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phenolsulfonphthalein Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8

Specifications

Melting Point >300°C
Molecular Formula (C6H4OH)2C7H4SO3
MDL Number MFCD00003552
Synonym Phenol red, Phenol red
Quantity 100 g
Molecular Weight (g/mol) 354.38
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.