missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, Supelco™

Product Code. 15877560
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877560 100 g 100g
15854960 5 g 5g
15867560 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15877560 Supplier Supelco Supplier No. 1072410100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator ACS

Phenol red (phenolsulfonphthalein, PH R) belongs to the class of triphenylmethane dyes. It is a waterͰsoluble anionic dye, mostly used as a pH indicator in cell biology laboratories. Also, in different protocols, it is used to detect H2O2(Hydrogen peroxide).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8

Specifications

Melting Point >300°C
Molecular Formula (C6H4OH)2C7H4SO3
MDL Number MFCD00003552
Synonym Phenol red, Phenol red
Quantity 100 g
Molecular Weight (g/mol) 354.38
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.