missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl Alcohol, TRC

Product Code. 30011847
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011847 1 g 1g
30135896 10 g 10g
30134996 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011847 Supplier Toronto Research Chemicals Supplier No. TRCP2960001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-12-8
Formula Weight 122.0732
InChI Formula InChI=1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2
IUPAC Name 2-phenylethanol
Molecular Formula C8 H10 O
Molecular Weight (g/mol) 122.16
Percent Purity >95%
SMILES OCCc1ccccc1
Synonym Phenethyl Alcohol, 2-Phenlylerthanol, Benzeneethanol, (2-Hydroxyethyl)benzene, β-(Hydroxyethyl)benzene, β-PEA, β-Phenethanol, β-Phenethyl alcohol, β-Phenylethanol, β-Phenylethyl alcohol, 2-Phenethanol, 2-Phenethyl alcohol, 2-Phenyl-1-ethanol, 2-Phenylethanol, 2-Phenylethyl alcohol, Benzyl carbinol, Ethanol, 2-phenyl-, NSC 406252, PEA, Phenethanol, Phenylethanol, Phenylethyl alcohol
Chemical Name or Material 2-Phenylethanol
Grade GC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.