Learn More
Phenazopyridine, TRC
Product Code. 30417965
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417965 1 g 1g
30413554 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30417965 Supplier Toronto Research Chemicals Supplier No. TRCP3137511G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 94-78-0
Molecular Formula C11H11N5
Molecular Weight (g/mol) 213.24
Synonym 3-(2-Phenyldiazenyl)-2,6-pyridinediamine Hydrochloride, 3-(Phenylazo)-2,6-pyridinediamine, 2,6-Diamino-3-phenylazopyridine, Gastracid, Gastrotest, NSC 145895
IUPAC Name 3-[(E)-phenyldiazenyl]pyridine-2,6-diamine
SMILES Nc1ccc(N=Nc2ccccc2)c(N)n1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C11H11N5/c12-10-7-6-9(11(13)14-10)16-15-8-4-2-1-3-5-8/h1-7H,(H4,12,13,14)
Formula Weight 213.1014
Chemical Name or Material Phenazopyridine