missing translation for 'onlineSavingsMsg'
Learn More

Phen-DC3 Trifluoromethanesulfonate, MedChemExpress

Product Code. p-200099090
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499217

Brand: Medchem Express HY15594A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phen-DC3 Trifluoromethanesulfonate is a G-quadruplex (G4) specific ligand which can inhibit FANCJ and DinG helicases with IC50s of 65±6 and 50±10 nM, respectively.

Phen-DC3 Trifluoromethanesulfonate is a G-quadruplex (G4) specific ligand which can inhibit FANCJ and DinG helicases with IC50s of 65±6 and 50±10 nM, respectively.

Specifications

Chemical Name or Material Phen-DC3 Trifluoromethanesulfonate
CAS 929895-45-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C36H26F6N6O8S2
Quantity 5 mg
Synonym Phen-DC3 Triflate
Solubility Information DMSO : ≥ 34 mg/mL (40.06 mM)
SMILES [O-]S(=O)(C(F)(F)F)=O.C[N+]1=C2C=CC=CC2=CC(NC(C3=CC=C4C=CC5=CC=C(C(NC6=C[N+](C)=C7C=CC=CC7=C6)=O)N=C5C4=N3)=O)=C1.[O-]S(=O)(C(F)(F)F)=O
Molecular Weight (g/mol) 848.75
Formula Weight 848.75
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.