missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phen-DC3 Trifluoromethanesulfonate, MedChemExpress
Description
Phen-DC3 Trifluoromethanesulfonate is a G-quadruplex (G4) specific ligand which can inhibit FANCJ and DinG helicases with IC50s of 65±6 and 50±10 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Phen-DC3 Trifluoromethanesulfonate |
| CAS | 929895-45-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H26F6N6O8S2 |
| Quantity | 10 mg |
| Synonym | Phen-DC3 Triflate |
| Solubility Information | DMSO : ≥ 34 mg/mL (40.06 mM) |
| SMILES | [O-]S(=O)(C(F)(F)F)=O.C[N+]1=C2C=CC=CC2=CC(NC(C3=CC=C4C=CC5=CC=C(C(NC6=C[N+](C)=C7C=CC=CC7=C6)=O)N=C5C4=N3)=O)=C1.[O-]S(=O)(C(F)(F)F)=O |
| Molecular Weight (g/mol) | 848.75 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?