missing translation for 'onlineSavingsMsg'
Learn More

Phen-DC3 Trifluoromethanesulfonate, MedChemExpress

Product Code. 16431539
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16431539 1 mg 1mg
16499217 5 mg 5mg
16496658 10 mg 10mg
16432747 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16431539

Brand: Medchem Express HY15594A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phen-DC3 Trifluoromethanesulfonate is a G-quadruplex (G4) specific ligand which can inhibit FANCJ and DinG helicases with IC50s of 65±6 and 50±10 nM, respectively.

Phen-DC3 Trifluoromethanesulfonate is a G-quadruplex (G4) specific ligand which can inhibit FANCJ and DinG helicases with IC50s of 65±6 and 50±10 nM, respectively.

Specifications

Chemical Name or Material Phen-DC3 Trifluoromethanesulfonate
CAS 929895-45-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C36H26F6N6O8S2
Quantity 1 mg
Synonym Phen-DC3 Triflate
Solubility Information DMSO : ≥ 34 mg/mL (40.06 mM)
SMILES [O-]S(=O)(C(F)(F)F)=O.C[N+]1=C2C=CC=CC2=CC(NC(C3=CC=C4C=CC5=CC=C(C(NC6=C[N+](C)=C7C=CC=CC7=C6)=O)N=C5C4=N3)=O)=C1.[O-]S(=O)(C(F)(F)F)=O
Molecular Weight (g/mol) 848.75
Formula Weight 848.75
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.