Learn More
Cytiva PhastGel™ Precast IEF Gels
Product Code. 10678225 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
pH 3 to 9
pH 4 to 6.5
pH 5 to 8
Unit Size:
Pack of 10
Product Code. pH Range unitSize
10678225 pH 3 to 9 Pack of 10
11768458 pH 5 to 8 Pack of 10
10546764 pH 4 to 6.5 Pack of 10
3 options
Product Code. 10678225
Brand: Cytiva 17054301
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Precast homogeneous gels containing Pharmalyte as carrier ampholyte to generate stable, linear pH gradients with uniform conductivity across the entire pH range and allow high field strengths for high resolution separations.
  • PhastGel IEF gels
  • Homogeneous gels in various pH ranges for separations of proteins by isoelectric point
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 125°C
Cassette Material Polyester
Description PhastGel IEF - 3-9
Gel Percentage 2 to 6%
Gel Thickness 0.45 mm
Gel Type Pharmalyte
pH Range pH 3 to 9
Separation of Protein
Separation Range pH 3 to 9
Separation Type Isoelectric Focusing
Formulation Containing 2 to 6% Pharmalyte as carrier Ampholyte
For Use With (Application) Charecterizing and Analysing Mixtures of Proteins with Smaller PI Differentials
Material GelBond polyester film
Melting Temperature 82° to 86°C
Molecular Weight (g/mol) 71.08
Quantity 10/Pk.
Storage Requirements 4 - 8°C
Thickness (English) 0.017 in.
Length (Metric) 50 mm
Length (Metric) Gel 4.6 mm
Width (English) 1.9 in.
Width (English) Gel 1.9 in.
Width (Metric) 48 mm
Width (Metric) Gel 5 cm
Height (English) 1.81 in.
Height (Metric) 4.6 cm
Product Type IEF Gel
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • EUH208-Contains Acrylamide. May produce an allergic reaction.
Precautionary Statement
  • P281-Use personal protective equipment as required.,Use personal protective equipment as required.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Acrylamide
  • UNKNOWN ACUTE-85 per cent of the mixture consists of component(s) of unknown acute toxicity.