Learn More
Ismatec™ PharMed™ BPT 2-stop Tubing
Product Code. 10083012
Click to view available options
Tubing Size:
2.79mm I.D., 4.49mm O.D.
Unit Size:
Pack of 6
Product Code. 10083012
Brand: Ismatec™ SC0746
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Formulated to withstand the rigors of peristaltic pumping action while providing the biocompatible fluid surface required in sensitive bioprocess applications. Ismatec™ PharMed™ BPT 2-stop Tubing requires extension tubing on both suction and discharge sides.
  • Outlasts silicone tubing in peristaltic pumps.
  • Withstands repeated autoclaving.
  • Withstands repeated CIP and SIP cleaning and sterilization.
  • Documented biocompatibility.
  • Multiple manufacturing sites.
  • Welded stoppers for autoclaving.
Specifications
Description Color-code: Purple-White
Autoclavable Autoclavable
Material PharMed BPT
Overall Length (Metric) 40 cm
Thickness (Metric) 0.85 mm
For Use With (Application) CA cassettes
Packaging Quantity 6 pcs./Pk.
Tubing Size 2.79mm I.D., 4.49mm O.D.
Product Type 2-stop Tubing