Description
- Co-polymerisates of glycine, glycylglycine, amines and epichlorhydrin
- Form extremely stable, linear pH gradients under the influence of electric fields
- Available in both narrow and broad pH ranges to suit specific application
- Broad ranges can be used directly for agarose, Ultrodex or polyacrylamide gels
- Narrow ranges provide high resolution for dedicated applications using polyacrylamide gels
Specifications
Specifications
| Color | Colorless to Light Yellow |
| pH Range | 4 to 6.5 |
| Form | Liquid |
| Formulation | Co-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin |
| For Use With (Application) | IEF |
| Quantity | 25mL |
| Storage Requirements | 2° to 8°C |
| Product Type | Pharmalyte Carrier Ampholyte |
| For Use With (Equipment) | PhastSystem Automated Electrophoresis Unit |