Learn More
Köttermann Pharmacy Fume Hood without Media
Product Code. 17352179
Change view
Click to view available options
Configuration Options:
No Ventilation Fan
Ventilation Fan
Plug Type:
GB Plug
Plug Type D (Old British Plug)
Schuko Plug
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Options Plug Type unitSize
17352179 No Ventilation Fan Plug Type D (Old British Plug) Each
17392179 No Ventilation Fan Schuko Plug Each
17332189 No Ventilation Fan GB Plug Each
17342179 Ventilation Fan Plug Type D (Old British Plug) Each
17382179 Ventilation Fan Schuko Plug Each
17322189 Ventilation Fan GB Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17352179 Brand Köttermann Brand No. 45100015
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Pharmacy Fume Hood without Media is designed for the handling of hazardous substances in pharmacies and is ideally suited for working with dusty preparations or volatile chemicals.
  • For working safely with dust-forming preparations and volatile chemicals
  • Long-lasting steel construction with one-piece polyacrylic sash
  • Ergonomic features to improve workflow
  • Low, compact design for small laboratories
Specifications
Configuration Options No Ventilation Fan
Dimensions (L x W x H) 640 x 900 x 1350 mm
Plug Type Plug Type D (Old British Plug)
Voltage 230 V
Weight (Metric) 115 kg