missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pharma Metals Calibration Standard 1: B, Ba, Cu, Fe, Zn @ 100; Ag, Al, As, Cd, Co, Cr, Li, Mn, Mo, Ni, Pb, Sb, Se, Sn, Tl, V @ 20 μg/mL in 2% HNO3/tr. HF, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Pharma Metals Calibration Standard 1: B, Ba, Cu, Fe, Zn @ 100; Ag, Al, As, Cd, Co, Cr, Li, Mn, Mo, Ni, Pb, Sb, Se, Sn, Tl, V @ 20 μg/mL in 2% HNO3/tr. HF
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Arsenic/Cadmium/Chromium/Cobalt/Lead/Lithium/Manganese/Molybdenum/Nickel/Selenium/Silver/Thallium/Tin/Vanadium/Barium/Boron/Copper/Iron/Zinc |
| CAS | 7429-90-5, 7440-36-0, 7440-38-2, 7440-43-9, 7440-47-3, 7440-48-4 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Antimony/Arsenic/Cadmium/Chromium/Cobalt/Lead/Lithium/Manganese/Molybdenum/Nickel/Selenium/Silver/Thallium/Tin/Vanadium/Barium/Boron/Copper/Iron/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 20 μg/mL/Antimony 20 μg/mL/Arsenic 20 μg/mL/Cadmium 20 μg/mL/Chromium 20 μg/mL/Cobalt 20 μg/mL/Lead 20 μg/mL/Lithium 20 μg/mL/Manganese 20 μg/mL/Molybdenum 20 μg/mL/Nickel 20 μg/mL/Selenium 20 μg/mL/Silver 20 μg/mL/Thallium 20 μg/mL/Tin 20 μg/mL/Vanadium 20 μg/mL/Barium 100 μg/mL/Boron 100 μg/mL/Copper 100 μg/mL/Iron 100 μg/mL/Zinc 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction