Learn More
Christeyns™ Phago'Spore Disinfectant Detergent
Product Code. 17870265
Change view
Click to view available options
Size:
750 mL
5 L
Unit Size:
10 L
4.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17870265 5 L 10 L
17860255 750 mL 4.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17870265 Supplier Christeyns™ Supplier No. C65007
  • ONLINE EXCLUSIVE PRICE
    1150.00 NOK
    1007.87 NOK / 10 L
    Save 142.13 NOK
    12% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Phago'Spore is a disinfectant detergent product for all surfaces, including the surfaces of non-invasive medical devices. Phago'Spore is formulated from an active substance (peracetic acid), guaranteeing broad-spectrum microbiological efficacy while ensuring respect for the environment through its biodegradability into acetic acid, oxygen and water.
  • Ready to use
  • Bactericidal, fungicidal, virucidal and sporicidal (active in 10 min. on C. difficile and in 5 min. on Norovirus)
  • TP02, TP04 biocide
  • Eco-friendly (active substance: peracetic acid)
  • 'Food contact' product (in accordance with the decree of September 8, 1999) if used with rinsing
  • Dense foam spray head to prevent aerosol formation and protect the user
Specifications
Product Type Sporicide/Disinfectant
Color-coded No
Form Clear Liquid
pH 2.1 to 2.3
Scent Fresh Scent
Sterility Non-sterile
Quantity 2 Cans
Size 5 L