Learn More
Christeyns™ Phago'Gel SPS
Product Code. p-200000166
Click to view available options
Volume (Metric):
100 mL
300 mL
1000 mL
5 L
Quantity:
12 Airless Bottles with 3 mL Pump
2 Cans
20 Bottles with 3 mL Pump
30 Bottles
Unit Size:
100 L
12 L
3 L
6 L
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17800275
Brand: Christeyns™ P65024
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Phago'Gel SPS is intended for the hygienic surgical disinfection of hands by friction as an alternative to simple or hygienic hand washing, in the absence of water, during serial contact, or before any aseptic gesture.
  • High tolerance
  • Fast to use
  • Bactericidal, fungicidal, mycobactericidal and virucidal
  • Fragrance-free, dye-free, phenoxyethanol-free and paraben-free
Specifications
Product Type Alcohol-based Hand Sanitizer Gel
Color Colorless
Form Gel
Quantity 30 Bottles
Volume (Metric) 100 mL