Learn More
Macherey-Nagel™ pH Paper
Product Code. 10549641 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
3.8 to 5.8
4 to 7
5.4 to 7
5.5 to 9
6.4 to 8
7.2 to 9.7
9 to 13
Unit Size:
5m
Each
Pack of 5
Product Code. pH Range unitSize
10549641 5.4 to 7 Pack of 5
10797941 1 to 14 5m
10098250 0.5 to 5.5 Each
10464491 1 to 11 Each
10323361 3.8 to 5.8 5m
10539641 4 to 7 Pack of 5
10559641 5.5 to 9 5m
10570022 6.4 to 8 5m
10613141 7.2 to 9.7 5m
10527341 9 to 13 Pack of 5
10 options
Product Code. 10549641
Brand: Macherey-Nagel™ 90208
Only null left
Add to basket
Add to basket
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
  • Universal and special indicator papers - simple pH measurement
  • Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
  • Always ready for use .
  • The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
Certifications/Compliance ISO 9001
pH Range 5.4 to 7
Format Roll
Graduations <5.4, 5.4, 5.7, 6.0, 6.2, 6.4, 6.7, 7.0 >7.0
Quantity 1 Each
Type pH Paper