Learn More
Hanna Instruments™ pH/mV Benchtop Meter
Product Code. 10209818
Change view
Click to view available options
Dimensions (L x W x H):
240 x 187 x 74mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10209818 240 x 187 x 74mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10209818 Supplier Hanna Instruments™ Supplier No. HI221102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High specification pH meter offering outstanding value for money.
  • High specification pH meter offering outstanding value for money
  • Designed with an extra wide LCD screen which can display pH/mV and temperature readings
  • Readings are automatically compensated for temperature to ensure accuracy
  • High specification and accuracy
  • pH/temperature or pH/mV and temperature versions
  • One or two calibration points
  • Up to five memorised calibration values
  • Supplied with refillable glass pH electrode, temperature probe, electrode stand, calibration sachets, electrode refill solution and mains power lead
Specifications
Accuracy (mV) ±0.2/±1mV
Accuracy (pH) ±0.01
Calibration Points Automatic, one or two-point with five memorized buffer values
Calibration Points (pH) Automatic, one or two-point with five memorized buffer values
Detection Range –2 to 16.00°C
Display Type LCD
Range (mV) ±399.9 mV, ±2000 mV
Range (pH) -2 to 16.00
Resolution (pH) 0.01
Temperature Range (Metric) -9.9°C to +120.0°C
Inputs BNC
Model HI2211-02
Power Supply 12 VDC
Product Type pH/mV Benchtop Meter
Resolution (mV) 0.1/1mV
Temperature Accuracy ±0.5°C
Temperature Compensation –9.9 to 120.0°C
Temperature (Metric) Operating -9.9°C to +120.0°C
Temperature Probe Calibration Stainless Steel temperature probe
Dimensions (L x W x H) 240 x 187 x 74mm
Voltage 230 V
Frequency 50Hz