Learn More
Macherey-Nagel™ pH-Fix pH Indicator Strips
Product Code. 15324194 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
0 to 14
1.7 to 3.8
2 to 9
Unit Size:
100 units
Pack of 100
Product Code. pH Range unitSize
15324194 1.7 to 3.8 100 units
15705802 0 to 14 Pack of 100
15364184 2 to 9 100 units
3 options
Product Code. 15324194
Brand: Macherey-Nagel™ 92190
Highly appreciated standard strips for many pH testing applications.
  • Easy dip and read procedure provides reliable results in seconds
  • Due to the patented pH-Fix technology, the indicator is chemically bound to the test pad
  • Dye does not wash out, so clothes stay clean and samples remain pure
Specifications
pH Range 1.7 to 3.8
Quantity 100 Strips
Type pH Indicator Paper
Length (Metric) 85 mm
Width (Metric) 6 mm