Learn More
Jenway™ pH Electrodes
Description
- Quickly reach equilibrium
- Significantly reduced response times
- Far greater reproducibility
- Significantly reduced long-term drift
Specifications
Specifications
| Connector Types | BNC |
| Diameter (Metric) | 12 mm |
| Type | Refillable Environmental Combination with Bulb End |
| Dimensions (L x W x H) | 12 dia. x 120 mm L |
| For Use With (Application) | measurements at low temperatures or in solutions of low ionic strength, general purpose pH measurement and accuratae Q.C. or research work |
| Length (Metric) | 120 mm |
| Material | Glass |
| pH Range | 0 to 11 |
| Temperature (Metric) Range | 0°C to 50°C |