Learn More
pH 5.5 Monobasic/Dibasic Potassium Phosphate Buffered Eluent in Accordance with United States Pharmacopoeia (USP), Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Buffered Eluent for Chromatography, Buffered Eluent Mobile Phase
- Developed, formulated, and specified in accordance with United States pharmacopeia requirements
- NIST traceable
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Item Description | pH 5.5 ± 0.02 pH units |
| Quantity | 500 mL |
| Storage | HDPE Bottle GL45 |
| For Use With | Mobile Phase Eluent |