Learn More
Schmidlin™ PG Plus FID Tower H2 and Zero Air Generator
This dual gas generator produces a continuous supply of high purity hydrogen and zero air.
Brand:  Schmidlin™ DB-FTHG500-EU
Additional Details : Weight : 24.00000kg
 View more versions of this product
Product Code. 17718889
  • 220045.21 NOK / Each
Estimated Shipment: 31-07-2024
Description
  • Increase efficiency and improve performance
  • On-demand supply 24/7
  • Zero air <0.1 ppm
  • THC Pressure: 11 barg (160 psig)
  • Permeation membrane drying system
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
Specifications
Specifications
58 cm
49 cm
11 bar
140 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
180 W
500 mL/min.
35°C
55 in.
Outlet: 1/8 in.
>99.9996%