Learn More
Abnova™ PFN1 (Human) Recombinant Protein
Product Code. p-7164554
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16128981
Brand: Abnova™ H00005216P01.10ug
Human PFN1 full-length ORF ( AAH06768, 1 a.a. - 140 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MAGWNAYIDNLMADGTCQDAAIVGYKDSPSVWAAVPGKTFVNITPAEVGVLVGKDRSSFYVNGLTLGGQKCSVIRDSLLQDGEFSMDLRTKSTGGAPTFNVTVTKTDKTLVLLMGKEGVHGGLINKKCYEMASHLRRSQY
Specifications
Accession Number AAH06768
Gene ID (Entrez) 5216
Name profilin 1
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Symbol PFN1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer