Learn More
PFK-015, MedChemExpress
Product Code. 16495766
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16495766
Brand: Medchem Express HY12204
MedChemExpress PFK-015 is an effective inhibitor of PFKFB3 with IC50 of 110 nM (recombinant PFKFB3) and inhibits PFKFB3 activity in cancer cells with IC50 of 20 nM.
PFK-015 is an effective inhibitor of PFKFB3 with IC50 of 110 nM (recombinant PFKFB3) and inhibits PFKFB3 activity in cancer cells with IC50 of 20 nM.
Specifications
Chemical Name or Material PFK-015
Physical Form Solid
CAS 4382-63-2
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H12N2O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 25.2 mg/mL (96.82 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=NC=C1)/C=C/C2=NC3=CC=CC=C3C=C2
Molecular Weight (g/mol) 260.29
Formula Weight 260.29
Percent Purity 98.29%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.