Learn More
Masterflex™ PFA Threaded Adapter
Description
- Available in a variety of materials and configurations to meet a wide range of laboratory needs
- Adapts from a NPT male thread to a hose barb.
Specifications
Specifications
| Description | Barb x NPT(M) Connection |
| Autoclavable | Autoclavable |
| Tubing Size | 1/4 in. |
| Hose Barb | 1/4 in. |
| Outlet Port | 1/4 in. |
| Port Size | 1/4 in. |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Shape | Straight |
| Product Type | NPT Male Adapter to Straight Barb |