Learn More
BrandTech™ VITALAB™ PFA Low-Form Griffin Beakers
Product Code. p-8004384
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Height (Metric):
50 mm
59 mm
72 mm
96 mm
122 mm
141 mm
Unit Size:
Each
Product Code. 10298820
Brand: BrandTech™ 110205
Only null left
Add to basket
Add to basket
These Low-Form Griffin Beakers have molded graduations that are easy to read and are resistant to wear from repeated use.
  • Nested design to conserve bench and storage space
  • Can withstand temperatures from −200° to +250°C
  • Autoclavable at 121°C and easy to clean
  • Superior chemical resistance and excellent long-term stability
Specifications
Color Transparent
Capacity (Metric) 25 mL
Material PFA
For Use With (Application) For Volume Measurement
Height (Metric) 50 mm
Quantity 1/Pk.
Temperature (Metric) Max. Operating +250°C
Type Measuring Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance DIN EN 285
Scale Raised Scale