missing translation for 'onlineSavingsMsg'
Learn More

PF-562271 besylate, MedChemExpress

Product Code. p-200096977
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490337

Brand: Medchem Express HY10458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-562271 (VS-6062) besylate is a potent ATP-competitive, reversible inhibitor of FAK and Pyk2 kinase, with an IC50 of 1.5 nM and 13 nM, respectively.

PF-562271 (VS-6062) besylate is a potent ATP-competitive, reversible inhibitor of FAK and Pyk2 kinase, with an IC50 of 1.5 nM and 13 nM, respectively.

Specifications

Chemical Name or Material PF-562271 besylate
CAS 939791-38-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H26F3N7O6S2
Quantity 50 mg
Synonym VS-6062 besylate
Solubility Information DMSO : 21.4 mg/mL (32.15 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CS(=O)(N(C)C1=NC=CC=C1CNC2=NC(NC3=CC4=C(NC(C4)=O)C=C3)=NC=C2C(F)(F)F)=O.O=S(C5=CC=CC=C5)(O)=O
Molecular Weight (g/mol) 665.66
Formula Weight 665.66
Percent Purity 95.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.