Learn More
PF-562271 besylate, MedChemExpress
Product Code. 16490337
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16490337
Brand: Medchem Express HY10458
MedChemExpress PF-562271 (VS-6062) besylate is a potent ATP-competitive, reversible inhibitor of FAK and Pyk2 kinase, with an IC50 of 1.5 nM and 13 nM, respectively.
PF-562271 (VS-6062) besylate is a potent ATP-competitive, reversible inhibitor of FAK and Pyk2 kinase, with an IC50 of 1.5 nM and 13 nM, respectively.
Specifications
Chemical Name or Material PF-562271 besylate
CAS 939791-38-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H26F3N7O6S2
Quantity 50 mg
Synonym VS-6062 besylate
Solubility Information DMSO : 21.4 mg/mL (32.15 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CS(=O)(N(C)C1=NC=CC=C1CNC2=NC(NC3=CC4=C(NC(C4)=O)C=C3)=NC=C2C(F)(F)F)=O.O=S(C5=CC=CC=C5)(O)=O
Molecular Weight (g/mol) 665.66
Formula Weight 665.66
Percent Purity 95.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.