missing translation for 'onlineSavingsMsg'
Learn More

PF-4840154, MedChemExpress

Product Code. p-200098001
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16443647 50 mg 50mg
16427638 10 mg 10mg
16417546 100 mg 100mg
16400108 5 mg 5mg
4 options
This item is not returnable. View return policy

Product Code. 16443647

Brand: Medchem Express HY18779

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-4840154 is a potent, selective agonist of the rat and human TrpA1 channel with EC50s of 97 and 23 nM, respectively. PF-4840154 elicits TrpA1-mediated nocifensive behaviour in mouse.

PF-4840154 is a potent, selective agonist of the rat and human TrpA1 channel with EC50s of 97 and 23 nM, respectively. PF-4840154 elicits TrpA1-mediated nocifensive behaviour in mouse.

Specifications

Chemical Name or Material PF-4840154
CAS 1332708-14-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H38N6O2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (107.15 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(N2CCN(CC3COCCC3)CC2)N=C1NCC(C)C)NCC4=CC=CC=C4
Molecular Weight (g/mol) 466.62
Formula Weight 466.62
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.