Learn More
PF-05085727, MedChemExpress
Product Code. 16476437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16476437
Brand: Medchem Express HY102050
MedChemExpress PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.
PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.
Specifications
Chemical Name or Material PF-05085727
CAS 1415637-72-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H18F3N7
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (151.19 mM; Need ultrasonic)
SMILES CN1C2=NC=NC(N3CCC3)=C2C(C4=C(C5=CC=C(C(F)(F)F)C=C5)N(C)N=C4)=N1
Molecular Weight (g/mol) 413.4
Formula Weight 413.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.