missing translation for 'onlineSavingsMsg'
Learn More

PF-04217903 methanesulfonate, MedChemExpress

Product Code. p-200098375
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497667

Brand: Medchem Express HY12017A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-04217903 methanesulfonate is a potent ATP-competitive c-Met kinase inhibitor with Ki of 4.8 nM for human c-Met. PF-04217903 methanesulfonate shows more than 1,000-fold selectivity relative to 208 kinases. Antiangiogenic properties.

PF-04217903 methanesulfonate is a potent ATP-competitive c-Met kinase inhibitor with Ki of 4.8 nM for human c-Met. PF-04217903 methanesulfonate shows more than 1,000-fold selectivity relative to 208 kinases. Antiangiogenic properties.

Specifications

Chemical Name or Material PF-04217903 methanesulfonate
CAS 956906-93-7
Color White
Purity Grade Notes Research
Molecular Formula C20H20N8O4S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (106.73 mM; Need ultrasonic)
SMILES OCCN1N=CC(C2=NC3=C(N=C2)N=NN3CC4=CC5=CC=CN=C5C=C4)=C1.CS(=O)(O)=O
Molecular Weight (g/mol) 468.49
Formula Weight 468.49
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.